Learn More
Integrin beta 8 Antibody (CL7290) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76480IR
Product Code. 30292701
  • 737.00 € / 0.10mL
Description
Integrin beta 8 Monoclonal antibody specifically detects Integrin beta 8 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Integrin beta 8
Monoclonal
DyLight 755
integrin beta-8, integrin, beta 8
This antibody was developed using a recombinant protein derived from P26012, with the exact immunogen sequence remaining proprietary.
0.1 mL
Signal Transduction
3696
Store at 4⪚C in the dark.
IgG1
Western Blot, Immunocytochemistry/Immunofluorescence
CL7290
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified