missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Interferon alpha-6 Monoclonal Antibody (1009720)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA560858
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
| Interferon alpha-6 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P05013 | |
| IFNA6 | |
| E. coli-derived recombinant human IFN-alpha 6/IFNA6, Leu28-Lys207. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 1009720 | |
| Unconjugated | |
| IFNA6 | |
| IFNA6; IFN-alpha-6; IFN-alphaK; Interferon; interferon alpha 6; interferon alpha-54; Interferon alpha-6; interferon alpha-K; interferon, alpha 6; leIF K; RP11-354P17.7 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3443 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction