missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interferon gamma Mouse, Clone: IFNG/466, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3458-MSM4-P0
This item is not returnable.
View return policy
Specifications
| Interferon gamma | |
| Cytoplasmic and Cell Surface | |
| IFNG/466 | |
| Unconjugated | |
| NP_000610 | |
| Mouse | |
| 20-25kDa | |
| Protein A/G Purified | |
| RUO | |
| 3458 | |
| IgG2b κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| IFNG | |
| Purified recombinant human IFN-gamma | |
| PMA stimulated human peripheral blood mononuclear cells. Renal or testicular carcinoma. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction