missing translation for 'onlineSavingsMsg'
Learn More

Internal Standard, SPEX CertiPrep™

Product Code. 12769705 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
2 mL
Unit Size:
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12769705 1 mL 1mL
12779705 2 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12769705 Supplier SPEX Certiprep Supplier No. CLPSI
  • ONLINE EXCLUSIVE PRICE
    139.52 €
    121.53 € / 1mL
    Save 17.99 €
    12% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,4-Dichlorobenzene-d43855-82-14000μg/mL
Acenaphthene-d1015067-26-24000μg/mL
Chrysene-d121719-03-54000μg/mL
Naphthalene-d81146-65-24000μg/mL
Perylene-d121520-96-34000μg/mL
Phenanthrene-d101517-22-24000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Internal Standard
Name Note EPA Method 625, SW-846 Method 8270C, CLP Methods
Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Methods For US EPA Methods 625, 8270C, CLP
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solvent or Matrix Methylene Chloride
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 4000μg/mL

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.