missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Intestinal Alkaline Phosphatase Recombinant Rabbit Monoclonal Antibody (23GB3645), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
207.00 € - 562.00 €
Specifications
| Antigen | Intestinal Alkaline Phosphatase |
|---|---|
| Clone | 23GB3645 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088833
![]() |
Invitrogen™
MA553434 |
100 μL |
562.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088199
|
Invitrogen™
MA553435 |
20 μL |
207.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Intestinal Alkaline Phosphatase Recombinant Monoclonal antibody specifically detects Intestinal Alkaline Phosphatase in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Intestinal Alkaline Phosphatase | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P09923 | |
| 248 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3645 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2010001C14Rik; Akp2; Akp6; Alkaline phosphatase 1, intestinal, defined by SSR; alkaline phosphatase 6; alkaline phosphatase, intestinal; alkaline phosphomonoesterase; Alp1; ALPI; AP; EC 3.1.3.1; gIAP; glycerophosphatase; I Kasahara isozyme; IAP; IAP-1; IAP-I; intestinal alkaline phosphatase; intestinal alkaline phosphatase 1; intestinal alkaline phosphatase I IAP-I; intestinal alkaline phosphatase III; intestinal alkaline phosphatase IV; intestinal alkaline phosphatase V; intestinal-type alkaline phosphatase; intestinal-type alkaline phosphatase 1; Kasahara isozyme; PLAP-1; Regan isozyme; S51097 | |
| A synthetic peptide of human intestinal alkaline phosphatase (1-55AA). | |
| Primary | |
| -20°C | |
| ALPI |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title