missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MultiShot™ StripWell Mach1™ T1 Phage-Resistant Chemically Competent E. coli

Product Code. 10032732
Change view
Click to view available options
Quantity:
1 x 96 tube rack (50 μl/tube)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10032732 1 x 96 tube rack (50 μl/tube) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10032732 Supplier Invitrogen™ Supplier No. C869601
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MultiShot StripWell Mach1 T1R chemically competent E. coli cells are exceptionally fast-growing cloning competent cells packaged in a rack containing 12 strips of 8 tubes for increased productivity of bacterial transformations.

MultiShot StripWell Mach1 T1R chemically competent E. coli cells are exceptionally fast-growing cloning competent cells packaged in a rack containing 12 strips of 8 tubes for increased productivity of bacterial transformations. The Mach1 T1R strain is derived from a non-K-12, wild-type W E. coli strain (ATCC #9637) that was selected for its rapid growth rate, and genetically manipulated to possess the most useful cloning strain features.

All of our T1R strains have the tonA (also known as fhuA) deletion in their genotype that confers resistance to T1 and T5 phage. T1 bacteriophage spread rapidly and lyse E. coli hosts, which are commonly used for cloning and library construction. A resistant genotype offers added security to protect valuable clones and libraries. This is especially important for genome and sequencing centers where T1 phage infection would be catastrophic.

Accelerate cloning workflow using Mach1 T1R strain
MultiShot StripWell Mach1 T1R competent E. coli cells are fast growing chemically competent cells with a doubling time of approximately 50 minutes, with other popular strains averaging 70 minutes. Mach1-T1R colonies are clearly visible within 8 hours of plating the transformation mix (ampicillin selection), enabling you to plate and pick colonies the same day. You can also prepare plasmid DNA 4 hours after inoculating a single, overnight-grown colony in the selective media of choice (with any antibiotic). The Mach1 T1R strain is useful for many cloning applications and provides transformation efficiencies in the MultiShot StripWell format of >1 x 108 cfu/μg with plasmid DNA.

The Mach1 T1R strain was engineered to carry the tonA mutation that confers resistance to T1 and T5 phage infection. The tonA genotype offers added security to valuable clones and libraries. The Mach1 T1R strain has the lacZΔM15 genotype, allowing blue-white screening on plates containing either X-Gal or Bluo-Gal. It also carries the recA1398 mutation that helps reduce the rate of recombination while propagating plasmid DNA, the endA1 mutation that increases plasmid DNA yield and quantity, and hsdR restriction system deletion to allow genomic DNA cloning.

MultiShot StripWell Mach1 T1R Competent Cells offer:
• Transformation efficiencies in StripWell format of >1 x 108 cfu/μg plasmid DNA
• tonA (fhuA) genotype to confer resistance to T1 and T5 phage
• lacZ∆M15 for blue-white color screening of recombinants
• hsdR mutation for efficient transformation of unmethylated DNA from PCR applications
• recA1398 mutation for reduced occurrence of homologous recombination in cloned DNA
• endA1 mutation for increased plasmid yield and quality

MultiShot StripWell flexible format
Each StripWell of eight connected tubes is designed with the same single-tube, single-use features as our One Shot format tubes. This format allows all steps of the transformation protocol, up to plating, to take place in the same tube, thereby saving time and reducing chance of contamination. After addition of DNA, MultiShot StripWell competent cells can be transformed by heat-shock at 42°C using a water bath, all in the same tube.

The StripWell tubes can be cut to permit transformations in a single tube, eliminating wasted aliquots of cells and avoiding freeze-thaws that can result in reduced transformation efficiency.

Benifits of the MultiShot StripWell format include:
• Increased productivity—suitable for high throughput experimentation
• Maximal flexibility—transform as few as 1 or as many as 96 tubes from the StripWell rack
• Familiar convenience—add DNA directly to cells, heat shock, and recover in the same tube

Genotype
F– φ80lacZΔM15 ΔlacX74 hsdR(rK–, mK+) ΔrecA1398 endA1 tonA

Find the strain and format that fits your needs
• A variety of E. coli strains and formats of chemically competent and electrocompetent cells are available to meet transformation needs.
• For high-throughput transformation choose from our collection of MultiShot comp cells.
• We offer bacterial growth media in various formats.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA No
Transformation Efficiency Level High Efficiency (>1 x 109 cfu/μg)
Content And Storage • Mach1 T1 Phage-Resistant (T1R) Chemically Competent E. coli (1 x 96-stripwell plate at 50 μL/well); store at –80°C
• pUC19 DNA (1 x 50 μL at 10 pg/μL); store at –20°C
• S.O.C. Medium (2 x 15 mL); store at 4°C or room temperature

Antibiotic Resistance Bacterial No
Cloning Unstable DNA Not suitable for cloning unstable DNA
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility High
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) Yes
Species E. coli (wild-type W)
Format 8-well Strips
Product Line MultiShot
Quantity 1 x 96 tube rack (50 μl/tube)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.