missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAX Efficiency™ DH10B Competent Cells

Product Code. 10593423
Change view
Click to view available options
Quantity:
5 x 200 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10593423 5 x 200 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10593423 Supplier Invitrogen™ Supplier No. 18297010
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

MAX Efficiency DH10B Competent Cells: 5 vials, 200μL each (total of 1mL), pUC19 DNA (0.01μg/mL): 1 vial, 100μL, S.O.C. medium: 2 bottles, 6mL each

MAX Efficiency DH10B Competent Cells are highly efficient E. coli suitable for a variety of molecular biology applications. This strain is one of the most commonly used strains for everyday applications due to key strain features such as high transformation efficiency.

MAX Efficiency DH10B Competent Cells are highly efficient E. coli suitable for a variety of molecular biology applications. This strain is one of the most commonly used strains for everyday applications due to key strain features such as high transformation efficiency, making them ideal for generating plasmid libraries, an ability to take up and maintain large plasmids (seen with plasmids of 150 kb in size), and a lack of methylation-dependent restriction systems.

Benefits of MAX Efficiency DH10B Competent Cells
• Ability to maintain large DNA plasmids 
• Resistant to the effects of ligase and ligase buffer and tolerant of small amounts of undiluted ligation reactions 
• Suitable for the construction of cDNA or genomic DNA libraries using plasmid-derived vectors 

Universal strain for everyday applications
The presence of the mcrA genotypic marker and the deletion of mcrBC and mrr makes this strain suitable for cloning DNA that contains methylcytosine and methyladenine. Therefore, genomic DNA, both prokaryotic and eukaryotic, can be cloned efficiently in DH10B.

Versatile DH10B genotype features
• lacZ∆M15 for blue/white screening of recombinant clones 
• Elimination of mcrA, mcrBC, mrr, and hsdRMS restriction systems to allow construction of more representative genomic libraries 
• endA1 mutation to increase plasmid yield and quantity 

Genotype
F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 endA1 araD139 Δ(ara-leu)7697 galU galK λ–rpsL(StrR) nupG

Find the strain and format that fits your need 
• DH10B Cells are available in both electrocompetent and chemically competent formats.
• MegaX DH10B T1R Electrocomp Cells (Cat. No.C640003) have the highest transformation efficiency (>3 x 1010 cfu/μg plasmid DNA).
• Multishot formats are available for high throughput applications.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA Yes (mcrA)
Transformation Efficiency Level High Efficiency (>1 x 109 cfu/μg)
Content And Storage • MAX Efficiency DH10B Competent Cells (5 x 200 μL)
Store Competent Cells at –70°C

• pUC19 DNA (100 μL at 0.01 μg/mL)
Store pUC19 DNA at –20°C.

• S.O.C. Medium (2 x 6 mL)
Store S.O.C. Medium at 4°C or room temperature.

Antibiotic Resistance Bacterial Yes (Streptomycin)
Cloning Unstable DNA Not suitable for cloning unstable DNA
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility Low
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (K12)
Format Tube
Product Line MAX Efficiency
Quantity 5 x 200 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.