missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAX Efficiency™ Stbl2™ Competent Cells

Product Code. 10799374
Change view
Click to view available options
Quantity:
5 x 200 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10799374 5 x 200 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10799374 Supplier Invitrogen™ Supplier No. 10268019
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

MAX Efficiency Stbl2 Competent Cells: 5 vials, 200μL each (total of 1mL), pUC19 DNA (0.01μg/mL): 1 vial, 100μL, SOC Medium: 1 bottle, 6mL

MAX Efficiency Stbl2 Competent Cells are high-efficiency chemically competent cells specifically designed for cloning unstable inserts.

MAX Efficiency Stbl2 Competent Cells are high-efficiency chemically competent cells specifically designed for cloning unstable inserts. In addition to recA1, a unique set of genetic markers allow for stable cloning of direct repeat and retroviral sequences and tandem array genes. The Stbl2 strain was used for the stable propagation of a plasmid containing approximately 100 direct repeats of 32 bp as well as full-length infectious simian immunodeficiency virus (SIV) clone can be maintained. This strain has also been demonstrated to stably propagate plant virus sequences such as potato virus Y (PVY).

MAX Efficiency Stbl2 Competent Cells are a derivative of the JM109/J5 strain. Competent cells are prepared by a patented modification of the procedure of Hanahan. The mcrA mutation and the mcrBC-hsdRMS-mrr deletion allow for the cloning of methylated genomic sequences.

MAX Efficiency Stbl2 Competent Cells offer:
• >1 x 109 transformants/μg plasmid DNA for cloning rare sequences
• Elimination of mcrBC-hsdRMS-mrr and mutation of mcrA that allows cloning of methylated genomic sequences
• A unique genotype that stabilizes direct repeats and retroviral sequences such as SIV and HIV
• Greatly increased plasmid yield and quality due to endA1 mutation
• recA1 for reduced occurrence of nonspecific recombination in cloned DNA

Genotype
F- mcrA Δ(mcrBC-hsdRMS-mrr) recA1 endA1 lon gyrA96 thi supE44 relA1 λ- Δ(lac-proAB)

Note: For optimal performance, expression in S.O.C. Medium and incubation on antibiotic plates at 30°C is recommended.

Find the strain and format that fits your needs
• A variety of Stbl strains are available with different genetic background or suitable for electroporation.
• The Stbl3 strain is available in MultiShot format for high throughput applications.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA Yes (mcrB, mrr)
Transformation Efficiency Level High Efficiency (>1 x 109 cfu/μg)
Content And Storage • MAX Efficiency Stbl2 Competent Cells (5 x 200 μL); store at –80°C
• pUC19 DNA (100 μL at 0.01 μg/mL); store at –20°C
• S.O.C. Medium (6 mL); store at 4°C or room temperature

Antibiotic Resistance Bacterial No
Cloning Unstable DNA Yes (recA1)
Blue/White Screening No
High-throughput Compatibility Low
Plasmid May be used for plasmids >20 kb
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (K12)
Format Tube
Product Line MAX Efficiency
Quantity 5 x 200 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.