missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MultiShot™ TOP10 Chemically Competent E. coli

Product Code. 11510146
Change view
Click to view available options
Quantity:
5 x 96-well Plate (15 μL/Well)
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11510146 5 x 96-well Plate (15 μL/Well) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11510146 Supplier Invitrogen™ Supplier No. C40005
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

MultiShot TOP10 Chemically Competent E. coli: Five 96-well plates, 15μL per well, pUC19 DNA (10pg/μL): 1 vial, 50μL, S.O.C. medium: 5 bottles, 15mL each

MultiShot TOP10 Chemically Competent E. coli cells are derived from DH10B and are one of the most popular laboratory strains.

MultiShot TOP10 Chemically Competent E. coli cells are derived from DH10B and are one of the most popular laboratory strains. Packaged in five 96-well microtiter plates (15 μL aliquots) to simplify high-throughput workflows and provide transformation efficiencies of >1 x 108 cfu/μg control plasmid DNA, Multishot TOP10 is a versatile strain that can be used in multiple routine cloning applications.

Benefits of MultiShot TOP10 competent cells
• TOP10 strain is ideal for high-efficiency cloning and plasmid propagation
• Microtiter plate format design is convenient for automated high-throughput cloning
• mcrA mutation supports efficient transfection of methylated DNA
• lacZΔM15 allele permits rapid blue/white screening of transfectants
• endA1 mutation reduce endonuclease activity and improve plasmid DNA yields
• recA1 mutation helps reduce unwanted recombination

Easy to use for high-throughput transformations
MultiShot TOP10 Chemically Competent cells in 96-well plates are designed to facilitate high-throughput transformation. This format allows all steps of the transformation protocol, up to plating, to take place in the same well, saving time and reducing the chance of contamination. After the addition of DNA, MultiShot TOP10 Chemically Competent cells can be transformed by heat-shock at 42°C using a heat block or thermocycler in the same plate.

Using our TA Cloning and TOPO Cloning protocols, including controls, yields of 100–400 colonies per agar plate can be expected. 

Genotype
F-mcrA Δ(mrr-hsdRMS-mcrBC) Φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara, leu)7697 galU galK rpsL (StrR) endA1 nupG

Find the strain and format that fits your needs
• DH strains are available in chemically competent and electrocompetent cell formats.
• For high-throughput transformation options, choose from our collection of MultiShot formatted comp cells.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA Yes (mcrA)
Transformation Efficiency Level Medium Efficiency (1 x 108 to 1 x 109 cfu/μg)
Content And Storage • MultiShot™ TOP10 Chemically Competent E. coli (5 x 96-well plates at 15 μL per well)
Store Competent Cells at –80°C.

• pUC19 DNA (1 x 50 μL)
Store pUC19 DNA at –20°C.

• S.O.C. medium (5 x 15 mL/bottle)
Store S.O.C. Medium at 4°C or at room temperature
Antibiotic Resistance Bacterial Yes (Streptomycin)
Cloning Unstable DNA Not suitable for cloning unstable DNA
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility High
Plasmid High Copy Plasmid
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (K12)
Format 96-well Plate
Product Line MultiShot
Quantity 5 x 96-well Plate (15 μL/Well)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.