missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ One Shot™ BL21(DE3)pLysE Chemically Competent E. coli

Product Code. 10276442
Change view
Click to view available options
Quantity:
21 x 50 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10276442 21 x 50 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10276442 Supplier Invitrogen™ Supplier No. C656503
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

One Shot BL21(DE3)pLysE Chemically Competent E. coli cells are ideal for use with bacteriophage T7 promoter-based expression systems such as pRSET and pET.

One Shot BL21(DE3)pLysE Chemically Competent E. coli cells are ideal for use with bacteriophage T7 promoter-based expression systems such as pRSET and pET. The pLysE plasmid carried by the BL21(DE3)pLysE strain produces higher amounts of T7 lysozyme than pLysS to further reduce basal level expression of the gene of interest. Like pLysS, pLysE confers resistance to chloramphenicol (CamR) and contains the p15A origin for compatibility with plasmids containing the ColE1 or pMB1 origin (i.e. pUC- or pBR322- derived plasmids). Recombinant proteins that are nontoxic to E. coli are generally expressed at higher levels in BL21(DE3) cells as compared to the BL21(DE3)pLysE strain with tighter expression control. The BL21(DE3)pLysE strain is recommended for use when expressing toxic genes (alternatively, BL21-AI could be considered).

The BL21 (DE3) E. coli strain is one of the most popular host strains to produce recombinant proteins and adapted in research or industrial applications (e.g., first biopharmaceutical recombinant human insulin). This strain had been engineered to carry the λDE3 lysogen that contains the T7 RNA polymerase gene under control of the lacUV5 promoter. IPTG is required to induce expression of the T7 RNA polymerase cascade system that in turn transcribe the T7 promoter regulated target gene.

BL21(DE3) is a derivative of the E. coli B strain that does not contain the lon protease and is also deficient in the outer membrane protease OmpT. The lack of two key proteases reduces degradation of heterologous proteins expressed in the strains. Additionally, several other strain features make them especially suited for protein production, namely fast growth in minimal medium, a lower acetate production when grown in high glucose media, and the ability to reach high cell density.

One Shot BL21(DE3)pLysE Chemically Competent E. coli offer:
• Transformation efficiency of >1 x 107 cfu/μg control plasmid DNA
• Ion and OmpT protease deficiency reduces degradation of recombinant protein
• IPTG-inducible lacUV5 promoter controls expression of the T7 RNA polymerase
• pLysE (CamR) plasmid for reduced basal expression in uninduced cells
• hsdS mutation allows efficient transformation of unmethylated DNA
• Fast growth in minimal medium and ability to reach high cell density

BL21(DE3)pLysE E. coli are ideal for use with T7 promoter-based expression vectors. Like BL21(DE3)pLysS E. coli, BL21(DE3)pLysE cells carry the lambda DE3 lysogen. In addition, BL21(DE3)pLysE cells contain the pLysE plasmid, which constitutively expresses T7 lysozyme. T7 lysozyme reduces the basal expression of target genes by inhibiting T7 RNA polymerase. The pLysE plasmid in BL21(DE3)pLysE expresses higher levels of T7 lysozyme than the pLysS plasmid in BL21(DE3)pLysS. The BL21(DE3)pLysE strain provides tighter control of T7 RNA polymerase, which is necessary when the recombinant protein to be expressed is toxic.

Easy-to-use One Shot format
 
BL21(DE3)pLysE Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. The single-tube, single-use format allows all steps of the transformation protocol, up to plating, to take place in the same tube, thereby helping save time and prevent contamination.

Genotype
F–ompT hsdSB (rB–, mB–) gal dcm (DE3) pLysE(CamR)

Find the strain and format that you need
• We offer other E. coli strains for protein expression.
• For expression of toxic proteins, consider BL21-AI One Shot Chemically Competent E. coli (Cat. No. C607003).
• BL21 Star (DE3) and other strains are available in MultiShot formats for high throughput applications.
• We offer a variety of systems for the expression of recombinant proteins in E. coli, the Champion pET expression system provides the highest level of protein production available in any expression system.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality No
Cloning Methylated DNA No
Transformation Efficiency Level Subcloning Efficiency (1 x 106 to 1 x 107 cfu/μg)
Content And Storage • BL21 Star (DE3)pLysE Chemically Competent E. coli (21 x 50 μL); store at –80°C
• pUC19 DNA (1 x 50 μL); store at –20°C
• S.O.C. Medium (6 mL); store at 4°C or room temperature

Antibiotic Resistance Bacterial Yes (Chloramphenicol)
Blue/White Screening No
High-throughput Compatibility Low
Preparing Unmethylated DNA Yes (dcm)
Reduces Recombination No
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (B)
Promoter T7
Format Tube
Product Line One Shot
Quantity 21 x 50 μL
Toxic Proteins No
Improves RNA Stability Yes (pLysE)
Improves Protein Stability Yes (lon, ompT)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.