missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ One Shot™ INVαF' Chemically Competent E. coli

Product Code. 10524973
Change view
Click to view available options
Quantity:
21 x 50 μL/tube
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10524973 21 x 50 μL/tube Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10524973 Supplier Invitrogen™ Supplier No. C202003
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

One Shot INVαF' Chemically Competent E. coli are designed to stably replicate high-copy number plasmids.

One Shot INVαF´ Chemically Competent E. coli are designed to stably replicate high-copy number plasmids. They allow for the preparation of high quality plasmid DNA with less carry over contaminants as compared to other popular strains such as JM109 and can achieve a transformation efficiency of >1 x 108 cfu/μg supercoiled plasmid.

INVαF´ cells carry the F´ episome and so can be utilized to produce single-stranded DNA (ssDNA) from vectors that have an f1 origin of replication. This strain is related to DH strains and features similar genetic traits and benefits. The lacZΔM15 marker provides α-complementation of the ß-galactosidase gene from pUC or similar vectors and can be used for blue-white screening of colonies on bacterial plates containing Bluo-Gal or X-Gal. In addition to supporting blue/white screening, recA1 and endA1 mutations increase insert stability and improve the quality of plasmid DNA.

One Shot INVαF´ Chemically Competent E. coli cells offer:
• Transformation efficiencies of >1 x 108 cfu/μg
• Stability and exceptional quality of high-copy number plasmid DNAb
• Blue-white screening of recombinant clones (lacZΔM15)
• Reduction of homologous recombination of transformed plasmids (recA1)
• Increased quality of plasmid DNA preparations (endA1)
• Production and rescue of ssDNA from vectors carrying the f1 origin of replication (F´)

Note:
INVαF´ cells do not require IPTG to induce expression from the lac promoter.

Convenient and efficient format
One Shot INVαF´ Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. Each tube contains enough cells for one complete transformation, reducing freeze-thaw cycles and limiting money wasted on unused cells.

Genotype
F´ endA1 recA1 hsdR17 (rK–, mK+) supE44 thi-1 gyrA96 relA1 φ80lacZΔM15 Δ(lacZYA-argF)U169 λ–
Genetic marker descriptions

Find the strain and format that fit your needs
• A variety of DH strains are available in chemically competent and electrocompetent cell formats.
• We offer other strains for ssDNA production, such as TOP10F´ (Cat. No. C303003) or electrocompetent DH12S (Cat. No. 18312017).
• MultiShot formats are available for high throughput applications.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome Yes
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA Yes (hsd)
Transformation Efficiency Level Medium Efficiency (1 x 108 to 1 x 109 cfu/μg)
Content And Storage • One Shot INVαF' Chemically Competent E. coli (21 x 50 μL)
Store Competent Cells at –80°C.

• pUC19 plasmid (50 μL at 10 pg/μL)
Store pUC19 plasmid at –20°C.

• S.O.C. Medium (6 mL)
Store S.O.C. Medium at 4°C or room temperature.

Antibiotic Resistance Bacterial No
Cloning Unstable DNA Not suitable for cloning unstable DNA
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility Low
Plasmid High Copy Plasmid
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (K12)
Format Tube
Product Line One Shot
Quantity 21 x 50 μL/tube
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.