missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ One Shot™ OmniMAX™ 2 T1R Chemically Competent E. coli

Product Code. 10022512
Change view
Click to view available options
Quantity:
21 x 50 μL/tube
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10022512 21 x 50 μL/tube Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10022512 Supplier Invitrogen™ Supplier No. C854003
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

One Shot OmniMAX 2 T1R E. coli: 21 vials, 50μL each, pUC19 DNA (10pg/μL): 1 vial, 50μL, S.O.C. medium: 1 bottle, 6mL

The One Shot OmniMAX 2 T1R E. coli strain is an improved chemically competent cell line that can achieve extremely high transformation efficiency of >5 x 109 transformants/μg pUC19 DNA. It offers the highest transformation efficiency of any chemically competent cells in a One Shot format.

The One Shot OmniMAX 2 T1R E. coli strain is an improved chemically competent cell line that can achieve extremely high transformation efficiency of >5 x 109 transformants/μg pUC19 DNA. It offers the highest transformation efficiency of any chemically competent cells in a One Shot format. OmniMAX 2 T1R is a versatile strain, perfect to use in all cloning applications, including Gateway technology and TOPO PCR cloning.

OmniMAX 2 T1R Chemically Competent E. coli cells carry the tonA (also known as fhuA) mutation, which confers resistance to T1 and T5 phage infection. The tonA genotype offers added security to valuable clones and libraries. This strain lacks the E. coli K12 restriction systems (mcrA Δ(mrr hsdRMS-mcrBC)), allowing efficient transformation of highly methylated DNA. The OmniMAX 2 T1R strain has the lacZΔM15 genotype, allowing blue-white screening on plates containing either X-Gal or Bluo-Gal. It also carries the recA1 mutation, which helps reduce the rate of recombination while propagating plasmid DNA, and the endA1 mutation that increases plasmid DNA yield and quantity. The OmniMAX 2 T1R strain contains an F´ episome that can support cloning plasmids with an f1-like origin of replication (phagemids). In addition, the F´ episome carries the lacIq repressor for inducible expression from trc, tac, and lac promoters using IPTG. Importantly, the ccdA gene present on the F´ episome is deleted, allowing negative selection of Gateway vectors as the strain is sensitive to the ccdB gene product.

One Shot OmniMAX 2 T1R Chemically Competent E. coli offer:
• Highest transformation efficiencies of >5 x 109 cfu/μg 
• tonA (fhuA) genotype to confer resistance to T1 and T5 phage
• Δ(ccdAB) for sensitivity to the toxic effects of the ccdB gene product, allowing negative selection of vectors containing the ccdB gene (Gateway cloning system)
• Stable F´ episome that carries lacIq repressor for inducible expression from trc, tac, and lac promoters
• Construction of more representative genomic libraries due to the elimination of mcrA, mcrBC, mrr, and hsdRMS
• lacZΔM15 for blue-white color screening of recombinant clones 
• endA1 for cleaner DNA preparations and better results in downstream applications due to elimination of nonspecific digestion by Endonuclease I 
• recA1 for reduced occurrence of nonspecific recombination in cloned DNA

Easy-to-use One Shot format 
OmniMAX 2 T1R Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. The single-tube, single-use format allows all steps of the transformation protocol, up to plating, to take place in the same tube, thereby helping save time and prevent contamination.

Genotype
F´ [proAB+lacIqlacZΔM15 Tn10(TetR) Δ(ccdAB)] mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 Δ(lacZYA-argF)U169 endA1 recA1 supE44 thi-1 gyrA96 (NalR) relA1 tonA panD

Find the strain and format that fit your needs
• We offer other strains in chemically competent and electrocompetent cell formats to meet your specific needs.
• OmniMAX 2 T1R and other strains are available in MultiShot formats for high throughput applications.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome Yes
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA Yes (mcrA)
Transformation Efficiency Level High Efficiency (>1 x 109 cfu/μg)
Content And Storage • One Shot OmniMAX 2 T1R E. coli (21 x 50 μL)
Store Competent Cells at –80°C.

• pUC19 DNA (50 μL at 10 pg/μL)
Store pUC19 DNA at –20°C.

• S.O.C. Medium (6 mL)
Store S.O.C. Medium at 4°C or room temperature.

Antibiotic Resistance Bacterial Yes (Tetracycline)
Cloning Unstable DNA Not suitable for cloning unstable DNA
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility Low
Plasmid May be used for plasmids >20 kb
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) Yes
Species E. coli (K12)
Format Tube
Product Line One Shot
Quantity 21 x 50 μL/tube
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.