missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ One Shot™ PIR2 Chemically Competent E. coli

Product Code. 10544963
Change view
Click to view available options
Quantity:
11 x 50 μL/tube
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10544963 11 x 50 μL/tube Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10544963 Supplier Invitrogen™ Supplier No. C111110
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

One Shot PIR2 Chemically Competent E. coli cells are for use with vectors that contain the R6Kγ origin of replication (e.g., pUni/V5-His-TOPO for recombination with an Echo cloning system).

One Shot PIR2 Chemically Competent E. coli cells are for use with vectors that contain the R6Kγ origin of replication (e.g., pUni/V5-His-TOPO for recombination with an Echo cloning system). The pir gene encodes the replication protein π, which is required to replicate and maintain plasmids containing the R6Kγ origin. The PIR2 strain contains the wild-type pir gene that maintains the donor at ∼15 copies per cell. This strain is recommended for maintaining constructs that express toxic genes or for libraries. PIR2 is made chemically competent and can achieve high transformation efficiency of >1 x 108 cfu/μg supercoiled plasmid.

R6Kγ allows tight control and selection and has been used in various applications including genome engineering, mutagenesis studies, bacterial artificial chromosome (BAC) construction, high-capacity extrachromosomal vector systems, and suicide plasmid construction.

One Shot PIR2 Chemically Competent E. coli cells offer:
• Wild-type pir gene to maintain vectors that contain the R6Kγ ori at ∼15 copies per cell
• Transformation efficiencies of >1 x 108 cfu/μg
• endA1 mutation that enables increased plasmid DNA yield and quantity
• recA1 mutation that reduces the occurrence of unwanted recombination in cloned DNA
• hsdR for efficient transformation of unmethylated DNA from PCR applications

Convenient and efficient format
PIR2 Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. Each tube contains enough cells for one transformation, reducing freeze-thaw cycles and money wasted on unused cells.

Genotype
F–∆lac169 rpoS(am) robA1 creC510 hsdR514 endA recA1 uidA(∆MluI)::pir
Genetic marker descriptions

Find the strain and format that fit your needs
• Multiple DH strains are available in chemically competent and electrocompetent cell formats.
• We offer the PIR1 strain (Cat. No. C101010) that carries mutated pir gene and maintain vectors at ∼250 copies per cell.
• MultiShot formats are available for high throughput applications.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA No
Content And Storage • One Shot PIR2 Chemically Competent E. coli (11 x 50 μL)
Store Competent Cells at –80°C.

• pUC19 plasmid (50 μL at 10 pg/μL)
Store pUC19 plasmid at –20°C.

• S.O.C. Medium
Store S.O.C. Medium at 4°C or room temperature.

Antibiotic Resistance Bacterial No
Blue/White Screening No
High-throughput Compatibility Low
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli
Format Tube
Product Line One Shot
Quantity 11 x 50 μL/tube
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.