missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ One Shot™ TOP10 Chemically Competent E. coli

Product Code. 10368022
Change view
Click to view available options
Quantity:
11 x 50 μL
21 x 50 μL
42 x 50 μL
Unit Size:
10 reactions
20 reactions
40 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10368022 11 x 50 μL 10 reactions
10666493 21 x 50 μL 20 reactions
10358022 42 x 50 μL 40 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10368022 Supplier Invitrogen™ Supplier No. C404010
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

One Shot TOP10 Chemically Competent E. coli are ideal for high-efficiency cloning and plasmid DNA propagation and are provided at a transformation efficiency of 1 x 109 cfu/μg plasmid DNA.

One Shot TOP10 Chemically Competent E. coli are ideal for high-efficiency cloning and plasmid DNA propagation and are provided at a transformation efficiency of 1 x 109 cfu/μg plasmid DNA. These cells allow stable replication of high-copy number plasmids and are the same competent cells that come with many of our cloning kits. TOP10 E. coli cells are genetically similar to the DH10B strain and have been reported to be more resilient to stress conditions like osmotic shock and acidic pH stress.

One Shot TOP10 cells:
• Maximize cloning efficiency in a single-tube format
• Provide enhanced genomic DNA cloning capabilities

TOP10 Chemically Competent E. coli cells carry mutations in the methylation-dependent restriction system (mcrA, mcrBC, and mrr) allowing the cloning of both prokaryotic and eukaryotic genomic DNA, as well as efficient plasmid rescue from eukaryotic genomes. Similar to other DH strains, TOP10 has the lacZΔM15 genotype, providing for the option of blue-white screening on plates containing either X-Gal or Bluo-Gal. The inclusion of recA1 and endA1 mutations increase insert stability and improve the quality of plasmid DNA prepared from minipreps.

One Shot TOP10 Chemically Competent E. coli cells offer:
• Transformation efficiencies of >1 x 109 cfu/μg
• hsdR for efficient transformation of unmethylated DNA from PCR amplifications
• mcrA for efficient transformation of methylated DNA from genomic preparations
• lacZΔM15 for blue/white color screening of recombinant clones
• endA1 for cleaner DNA preparations and better results in downstream applications due to elimination of nonspecific digestion by Endonuclease I
• recA1 for reduced occurrence of nonspecific recombination in cloned DNA
• Expression from the lac promoter without IPTG

Easy-to-use One Shot format
TOP10 Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. The single-tube, single-use format accomodates allows all steps of the transformation protocol, up to plating, to take place in the same tube, helping save time and prevent contamination.

Genotype
F–mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697 galU galK λ–rpsL(StrR) endA1 nupG

Find the strain and format that fit your needs 
• We offer other DH strains in chemically competent and electrocompetent cell formats.
• The TOP10 strain is available in several MultiShot formats for high throughput applications.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality Yes (endA1)
Cloning Methylated DNA Yes (mcrA)
Transformation Efficiency Level High Efficiency (>1 x 109 cfu/μg)
Content And Storage • One Shot TOP10 Chemically Competent E. coli (11 x 50 μL)
Store Competent Cells at –80°C.

• pUC19 DNA (50 μL at 10 pg/μL)
Store pUC19 DNA at –20°C.

• S.O.C. medium (6 mL)
Store S.O.C. Medium at 4°C or room temperature.

Antibiotic Resistance Bacterial Yes (Streptomycin)
Cloning Unstable DNA No
Blue/White Screening Yes (lacZΔM15)
High-throughput Compatibility Low
Plasmid High Copy Plasmid
Preparing Unmethylated DNA No
Reduces Recombination Yes (recA1)
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (K12)
Format Tube
Product Line One Shot
Quantity 11 x 50 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.