missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UBE2L6 Polyclonal Antibody

Product Code. 16314015
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16314015 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16314015 Supplier Invitrogen™ Supplier No. PA587155
  • ONLINE EXCLUSIVE PRICE
    648.00 €
    460.08 € / 100µL
    Save 187.92 €
    29% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s) and ubiquitin-protein ligases (E3s). UBE2L6 is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is demonstrated to function in the E6/E6-ap-induced ubiquitination of p53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2L6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene UBE2L6
Gene Accession No. O14933, Q4V8J2, Q9QZU9
Gene Alias 2810489I21Rik; E2 ubiquitin conjugating enzyme; E2 ubiquitin-conjugating enzyme L6; MGC40331; retinoic acid induced gene B protein; retinoic acid-induced gene B protein; RIG-B; Ubce8; UBCH8; Ubcm8; UBE2L6; Ubiquitin carrier protein L6; ubiquitin conjugating enzyme E2 L6; ubiquitin conjugating enzyme E2L 6; Ubiquitin/ISG15-conjugating enzyme E2 L6; ubiquitin-conjugating enzyme 8; ubiquitin-conjugating enzyme E2L 6; Ubiquitin-protein ligase L6
Gene Symbols UBE2L6
Host Species Rabbit
Immunogen A synthetic peptide of human UBE2L6.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295704, 56791, 9246
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.