missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MC1061/P3 Chemically Competent E. coli

Product Code. 10388162
Change view
Click to view available options
Quantity:
5 x 300 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10388162 5 x 300 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10388162 Supplier Invitrogen™ Supplier No. C66303
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MC1061/P3 Chemically Competent E. coli are used for highly efficient transformations that require the P3 episome for selection and maintenance of vectors encoding the tyrosine tRNA suppressor (synthetic supF gene, such as pCDM8, pcDNA1.1, or any other supF-containing vector).

MC1061/P3 Chemically Competent E. coli are used for highly efficient transformations that require the P3 episome for selection and maintenance of vectors encoding the tyrosine tRNA suppressor (synthetic supF gene, such as pCDM8, pcDNA1.1, or any other supF-containing vector).

MC1061 E. coli cells were used by Hanahan and coworkers to engineer the DH10B strain. Among the replaced genes by adding mutations were recA1 (inhibiting the homologous recombination system), endA1, and lacZΔM15. Thus, this strain with its intact homologous recombination system is used in applications where recombination is needed, such as in genetically engineering E. coli by recombination or plasmid-phage recombination.

MC1061/P3 Chemically Competent E. coli offer:
• Transformation efficiencies of >1 x 108 cfu/μg
• P3 plasmid for selection of supF-containing plasmids
• hsdR for efficient transformation of unmethylated DNA from PCR amplifications

P3 plasmid
The P3 plasmid is a low-copy-number, 60 kb plasmid that carries kanamycin, tetracycline, and ampicillin resistance markers. The kanamycin gene is fully active and used to select for cells carrying P3. The tetracycline and ampicillin genes carry amber mutations, which render the genes inactive during normal growth and replication of the bacteria. Upon transformation of a vector carrying the suppressor F gene, the amber mutations in the tetracycline and ampicillin genes on the P3 plasmid are suppressed, and the E. coli are resistant to these antibiotics.

Note: The spontaneous reversion rate for the amber mutation in the ampicillin marker is 5%, while the reversion rate for the amber mutation in the tetracycline marker is 1%.To minimizing the reversion rates, use tetracycline and ampicillin together for selection of E. coli (P3) cells transformed with the SupF plasmid.

Genotype
F–hsdR(rK–, mK+) araD139 Δ(araABC-leu)7679 galU galK ΔlacX74 rpsL(StrR) thi mcrB / P3: KanR AmpR (am) TetR (am)

Find the strain and format that fits your needs
• We offer DH strains in chemically competent and electrocompetent cell formats to meet your specific needs.
• The TOP10/P3 strain (Cat. No. C505003) could be used where recombination is not needed.
• Strains are available in several MultiShot formats for high throughput applications.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality No
Cloning Methylated DNA Yes (hsd)
Transformation Efficiency Level Medium Efficiency (1 x 108 to 1 x 109 cfu/μg)
Content And Storage • MC1021/P3 Chemically Competent E. coli (5 x 300 μL)
Store Competent Cells at –80°C.

• pUC19 DNA (1 x 50 μL)
Store pUC19 DNA at –20°C.

• S.O.C. Medium (15 ml)
Store S.O.C. Medium at 4°C or room temperature.

Antibiotic Resistance Bacterial Yes (Streptomycin, Kanamycin, Ampicillin, Tetracycline)
Cloning Unstable DNA No
Blue/White Screening No
High-throughput Compatibility Low
Preparing Unmethylated DNA No
Reduces Recombination No
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (K12)
Format Tube
Quantity 5 x 300 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.