missing translation for 'onlineSavingsMsg'
Learn More

Iodide Standard For Ion Chromatography, SPEX CertiPrep™

Product Code. 11995074 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
Unit Size:
125mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11995074 125 mL Triple-leached LDPE bottle 125mL
11985074 500 mL Triple-leached LDPE bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11995074 Supplier SPEX Certiprep Supplier No. ASI92Y
  • ONLINE EXCLUSIVE PRICE
    97.98 €
    76.80 € / 125mL
    Save 21.18 €
    21% Off
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
I-1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7681-82-5 , 7732-18-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 99.9, 0.1
Analyte or Component Names Sodium iodide,distilled water,conductivity or of similar purity
Technique Ion Chromatography
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
Type Certified Reference Material
Element Sodium Iodide
Solvent or Matrix Water
Quantity 125 mL
Concentration or Composition (by Analyte or Components) 1000mg/L
Chemical Name or Material Iodide Standard, For Ion Chromatography
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.