missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Iodine, 0.01N Standardized Solution

Product Code. 11313767
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313767 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11313767 Supplier Thermo Scientific Chemicals Supplier No. 035636.K2
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7553-56-2 | I2 | 253.81 g/mol

Iodine, 0.01N standardized solution is used for accurate volumetric analysis. It is also used as a disinfectant and antibacterial agent in solutions, tinctures and salves.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iodine
CAS 7553-56-2, 7681-11-0, 7732-18-5
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Molecular Formula I2
MDL Number MFCD00011355 MFCD00164163
Color Red to Brown
Solubility Miscible with chloroform, carbon tetrachloride, carbon disulfide, cyclohexane, methanol, ethyl acetate, toluene, benzene, n-hexane, butan-2-ol, bromoethane, n-heptane, glycerol and diethyl ether
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Type Analytical Volumetric Solution
SMILES II
IUPAC Name diiodine
Molecular Weight (g/mol) 253.81
PubChem CID 807
Quantity 1 L
ChEBI CHEBI:17606
Formula Weight 253.81
Grade Standard
Concentration or Composition (by Analyte or Components) Iodine: 0.13%; Potassium iodide: 2.5%; Water: 97.37%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.