missing translation for 'onlineSavingsMsg'
Learn More

Iodine monochloride, 1M soln. in dichloromethane

Product Code. 15467945
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467945 500 mL 500mL
15457945 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15467945 Supplier Thermo Scientific Alfa Aesar Supplier No. H31938.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7790-99-0 | ClI | 162.35 g/mol

Iodine monochloride is a useful reagent in organic synthesis and source of I+. It is used as a source of electrophilic iodine in the synthesis of certain aromatic iodides. It also cleaves C-Si bonds. Iodine monochloride will also add to the double bond in alkenes to give chloro-iodo alkanes. It can be dissolved in acetic acid to make Wijs solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-99-0
Molecular Formula ClI
Molecular Weight (g/mol) 162.35
MDL Number MFCD00011354
InChI Key QZRGKCOWNLSUDK-UHFFFAOYSA-N
Synonym iodine monochloride, iodine chloride, iodochlorine, iodine chloride icl, chloroiodide, wijs' chloride, iodinemonochloride, chlorine iodide, unii-0smg5nlu45, protochlorure d'iode french
PubChem CID 24640
IUPAC Name iodochlorane
SMILES ClI

Specifications

Linear Formula ICl
UN Number UN2922
Concentration 1M soln. in dichloromethane
Density 1.42
Solubility Information Insoluble in water.
Formula Weight 162.36
Quantity 500 mL
Sensitivity Air and light sensitive
Chemical Name or Material Iodine monochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.