missing translation for 'onlineSavingsMsg'
Learn More

Iodipamide, TRC

Product Code. 30012308
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012308 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30012308 Supplier Toronto Research Chemicals Supplier No. TRCI6668005MG
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 606-17-7
Molecular Formula C20 H14 I6 N2 O6
Quantity 5 mg
Synonym 3,3'-[(1,6-dioxo-1,6-hexanediyl)diimino]bis[2,4,6-triiodo-benzoic acid, Benzoic acid, 3,3'-(adipoyldiimino)bis[2,4,6-triiodo- (6CI,8CI), 3,3'-(Adipoyldiimino)-bis(2,4,6-triiodobenzoic acid), Adipic acid bis(3-carboxy-2,4,6-triiodoanilide), Adipic acid di(3-carboxy-2,4,6-triiodoanilide), Adipiodon, Adipiodone, Bilignost, Biligrafin, Cholografin, Iodipamic acid, Iodipamide, N,N'-Adipylbis(3-amino-2,4,6-triiodobenzoic acid), Transbilix
InChI Formula InChI=1S/C20H14I6N2O6/c21-7-5-9(23)17(15(25)13(7)19(31)32)27-11(29)3-1-2-4-12(30)28-18-10(24)6-8(22)14(16(18)26)20(33)34/h5-6H,1-4H2,(H,27,29)(H,28,30)(H,31,32)(H,33,34)
SMILES OC(=O)c1c(I)cc(I)c(NC(=O)CCCCC(=O)Nc2c(I)cc(I)c(C(=O)O)c2I)c1I
IUPAC Name 3-[[6-(3-carboxy-2,4,6-triiodoanilino)-6-oxohexanoyl]amino]-2,4,6-triiodobenzoic acid
Molecular Weight (g/mol) 1139.76
Formula Weight 1139.512
Chemical Name or Material Iodipamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.