missing translation for 'onlineSavingsMsg'
Learn More

Iodobenzene, TRC

Product Code. 30406376
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406376 50 g -
30402504 1 g 1g
30389238 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406376 Supplier Toronto Research Chemicals Supplier No. TRCI68950050G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-50-4
Molecular Formula C6 H5 I
Molecular Weight (g/mol) 204.01
Synonym Benzene, iodo-, Iodobenzene, 1-Iodobenzene, 4-Iodobenzene, Benzene iodide, NSC 9244, Phenyl iodide
IUPAC Name iodobenzene
SMILES Ic1ccccc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H5I/c7-6-4-2-1-3-5-6/h1-5H
Formula Weight 203.9436
Chemical Name or Material Iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.