accessibility menu, dialog, popup

UserName

I

CAS RN 591-50-4

IUPAC Name: iodobenzene
Synonyms: iodophenyl
Molecular Weight (g/mol): 204.01
Molecular Formula: C6H5I
InChi Key: SNHMUERNLJLMHN-UHFFFAOYSA-N
SMILES: IC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)