missing translation for 'onlineSavingsMsg'
Learn More

Iodomethylcyclopentane, 98%, stabilized

Product Code. 10289972 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10289972 1 g Glass bottle 1g
10532603 5 g Glass bottle 5g
11924511 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10289972 Supplier Thermo Scientific Acros Supplier No. 378640010
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27935-87-1 | C6H11I | 210.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27935-87-1
Molecular Formula C6H11I
Molecular Weight (g/mol) 210.06
MDL Number MFCD03453190
InChI Key DUMSKQUKLVSSII-UHFFFAOYSA-N
Synonym iodomethyl cyclopentane, cyclopentylmethyl iodide, cyclopentane, iodomethyl, cyclopentyliodomethane, acmc-20akpz, cyclopentylmethyliodide, iodomethyl-cyclopentane, cyclopentanemethyl iodide
PubChem CID 2762531
IUPAC Name iodomethylcyclopentane
SMILES ICC1CCCC1

Specifications

Boiling Point 78.0°C (17.0 mmHg)
Color Brown or Colorless to Orange
Physical Form Solution
Quantity 1 g
Assay Percent Range 97.5% min. (GC)
Linear Formula (C5H9)CH2I
Flash Point 92°C
Solubility Information Solubility in water: immiscible
Formula Weight 210.05
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5400 to 1.5440
Specific Gravity 1.6
Density 1.6000g/mL
Chemical Name or Material Iodomethylcyclopentane, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • Iodomethylcyclopentane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.