missing translation for 'onlineSavingsMsg'
Learn More

Iodosobenzene diacetate, 98+%

Product Code. 11420368
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11420368

Brand: Thermo Scientific Alfa Aesar undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3240-34-4 | C10H11IO4 | 322.098 g/mol

Iodosobenzene is used as an oxidizing and acetoxylating agent in organic synthesis. It is actively involved in the preparation of (Z)-3,7-dimethyl-2,6-octadien-1-al(neral) from (Z)-3,7-dimethyl-2,6-octadien-1-ol (nerol) in presence of 2,2,6,6-tetramethylpiperidin-1-oxyl (TEMPO). It is a useful reagent for the synthesis of a wide variety of heterocyclic compounds. It is also used in the Pd-catalyzed 2-arylation of indoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodosobenzene is used as an oxidizing and acetoxylating agent in organic synthesis. It is actively involved in the preparation of (Z)-3,7-dimethyl-2,6-octadien-1-al(neral) from (Z)-3,7-dimethyl-2,6-octadien-1-ol (nerol) in presence of 2,2,6,6-tetramethylpiperidin-1-oxyl (TEMPO). It is a useful reagent for the synthesis of a wide variety of heterocyclic compounds. It is also used in the Pd-catalyzed 2-arylation of indoles.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3240-34-4
Molecular Formula C10H11IO4
Molecular Weight (g/mol) 322.098
MDL Number MFCD00008692
InChI Key ZBIKORITPGTTGI-UHFFFAOYSA-N
Synonym iodobenzene diacetate, diacetoxyiodo benzene, iodosobenzene diacetate, phenyliodine diacetate, phenyliodo diacetate, phenyliodoso acetate, phenyliodosodiacetate, phenyliodoso diacetate, bis acetato phenyliodine, benzene, diacetoxyiodo
PubChem CID 76724
IUPAC Name [acetyloxy(phenyl)-$l^{3}-iodanyl] acetate
SMILES CC(=O)OI(C1=CC=CC=C1)OC(=O)C

Specifications

Melting Point 159°C to 161°C
Quantity 100 g
Beilstein 1879369
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 322.1
Percent Purity ≥98%
Chemical Name or Material Iodosobenzene diacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.