missing translation for 'onlineSavingsMsg'
Learn More

Iohexol, MedChemExpress

Product Code. 16492176
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492176 500 mg 500mg
16428386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492176 Supplier Medchem Express Supplier No. HYB0594
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Iohexol is a radiographic contrast agent and can be applied for myelography, computerized tomography (cisternography, ventriculography) and MicroCT imaging in vivo.

Iohexol is a radiographic contrast agent and can be applied for myelography, computerized tomography (cisternography, ventriculography) and MicroCT imaging in vivo.

Specifications

Chemical Name or Material Iohexol
CAS 66108-95-0
Color White
Purity Grade Notes Research
Molecular Formula C19H26I3N3O9
Quantity 500 mg
Solubility Information H2O : ≥ 50 mg/mL (60.89 mM) ∣DMSO : 50 mg/mL (60.89 mM; Need ultrasonic)
SMILES O=C(C1=C(I)C(N(C(C)=O)CC(O)CO)=C(I)C(C(NCC(O)CO)=O)=C1I)NCC(O)CO
Molecular Weight (g/mol) 821.14
Formula Weight 821.14
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.