missing translation for 'onlineSavingsMsg'
Learn More

Potassium Single Element Ion Standard - Cation, SPEX CertiPrep™

Product Code. 11420199 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420199 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11420199 Supplier SPEX Certiprep Supplier No. CSK22Y

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
K1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 7757-79-1, 7697-37-2, 7732-18-5
Certificate of Analysis With Certificate of Analysis.
CAS Min % 0.1, 0.2, 99.7
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 0.1, 0.2, 99.7
Molecular Formula KNO3
MDL Number MFCD00011409
Technique Ion Chromatography
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Type Certified Reference Material
SMILES [K+].[O-][N+]([O-])=O
IUPAC Name potassium nitrate
Solvent or Matrix Nitric Acid
Molecular Weight (g/mol) 101.10
PubChem CID 24434
Quantity 125 mL
ChEBI CHEBI:63043
Chemical Name or Material Potassium Cation Standard
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.