missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate Eluent Concentrate For Ion Chromatography, 0.5M, SPEX CertiPrep™

Product Code. 15658295 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
This item is not returnable. View return policy

Product Code. 15658295

Brand: SPEX Certiprep ICELCON2100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
NaHCO30.5M

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 144-55-8, 7732-18-5
Certificate of Analysis With Certificate of Analysis
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Molecular Formula CHNaO3
MDL Number MFCD00003528
Technique Ion Chromatography
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Type Certified Reference Material
SMILES [Na+].OC([O-])=O
IUPAC Name sodium;hydrogen carbonate
Solvent or Matrix Water
Molecular Weight (g/mol) 84.01
PubChem CID 516892
Quantity 125 mL
ChEBI CHEBI:32139
Chemical Name or Material Sodium Bicarbonate Eluent Concentrate
Show More Show Less

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.