missing translation for 'onlineSavingsMsg'
Learn More

Melford Ionomycin

Product Code. p-200087081
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 30018353

Brand: Melford I520200.001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ionomycin free acid is frequently employed in cell biology experiments to study cellular processes that are dependent on calcium signaling.

Ionomycin free acid is frequently employed in cell biology experiments to study cellular processes that are dependent on calcium signaling. By transporting calcium ions into cells, ionomycin can trigger various cellular responses, including the activation of enzymes and signaling pathways. Researchers use ionomycin in a variety of applications, such as studying cell signaling, apoptosis, and calcium-dependent processes. It is particularly useful in experiments where precise control over intracellular calcium levels is necessary.

Specifications

Chemical Name or Material Ionomycin
CAS 56092-81-0
Quantity 1 mg
Molecular Formula C41H72O9
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.