missing translation for 'onlineSavingsMsg'
Learn More

Melford Ionomycin Calcium

Product Code. 30018356
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018356 5 mg 5mg
30018355 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018356 Supplier Melford Supplier No. I520250.005
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Ionomycin Calcium Salt is a calcium ionophore, which means it facilitates the transport of calcium ions across cellular membranes.

Ionomycin Calcium Salt is a calcium ionophore, which means it facilitates the transport of calcium ions across cellular membranes.

It is commonly employed to study calcium signaling and its role in various biological processes. It is particularly useful for investigating intracellular calcium levels, calcium-dependent enzymes, and cellular responses triggered by calcium influx.

Ionomycin Calcium Salt is used in cell culture experiments to induce a rapid and controlled increase in intracellular calcium levels. This allows the investigation of the downstream effects of calcium signaling pathways, including cellular activation, secretion, proliferation, and apoptosis.

Furthermore, it is frequently utilized in combination with other substances, such as fluorescent calcium indicators or protein kinase activators, to elucidate specific cellular mechanisms and signaling pathways.

Specifications

Chemical Name or Material Ionomycin, Calcium
CAS 56092-82-1
Quantity 5 mg
Molecular Formula C41H70O9Ca
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.