missing translation for 'onlineSavingsMsg'
Learn More

Ionomycin (Calcium Salt), Fisher BioReagents™

Product Code. 10429883
Change view
Click to view available options
Quantity:
1 mg
5 mg
Packaging:
Poly Micro Tube
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10429883 1 mg Poly Micro Tube 1mg
11438343 5 mg Poly Micro Tube 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10429883 Supplier Fisher BioReagents Supplier No. BP25271
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ionomycin (Calcium Salt)

  •  Suitable for use in electrophoretic separations, specifically polyacrylamide gel electrophoresis (PAGE)
  • This product is a 40% (w/v) solution of high quality acrylamide
  • Free of DNase, RNase, and protease
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56092-82-1
Molecular Formula C41H70CaO9
Molecular Weight (g/mol) 747.08
MDL Number MFCD00083634
InChI Key RULWQOBWZOQCOY-SQNLDGQHSA-M
PubChem CID 71311740
IUPAC Name calcium;(4R,6S,8S,10E,12R,14R,16E,18R,19R,20S,21S)-19,21-dihydroxy-22-[(2S,5S)-5-[(2R,5S)-5-[(1R)-1-hydroxyethyl]-5-methyloxolan-2-yl]-5-methyloxolan-2-yl]-4,6,8,12,14,18,20-heptamethyl-11-oxido-9-oxodocosa-10,16-dienoate
SMILES [Ca++].C[C@H](CCC([O-])=O)C[C@H](C)C[C@H](C)C(=O)[CH-]C(=O)[C@H](C)C[C@H](C)C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@@H](O)C[C@@H]1CC[C@](C)(O1)[C@H]1CC[C@](C)(O1)[C@@H](C)O

Specifications

Type Ionomycin Calcium Salt
Physical Form Solid
Melting Point 205°C to 206°C
Packaging Poly Micro Tube
Color White
Quantity 1 mg
Assay Percent Range ≥98.0 %
Formula Weight 747.1
Percent Purity 98.0% min. (by NP TLC), 99.0% min. (by RP HPLC)
Chemical Name or Material Ionomycin, Calcium Salt, From Streptomyces conglobatus
compliance-icons
Product Identifier
  • Ionomycin, Calcium Salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.