Learn More
Thermo Scientific IOPQ Package - CryoExtra
Product Code. 30503411
Change view
Click to view available options
Product Type:
IQ/OQ Service
Unit Size:
Each
Product Code. Product Type Unit Size
30503411 IQ/OQ Service Each
1 options
Product Code. 30503411 Supplier Thermo Scientific Supplier No. IOPQPCKE89003811
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Installation and Operational Qualification with temperature mapping and compliance documentation for CryoExtra systems.
  • Verifies that the equipment meets required specifications and functions correctly
  • Includes key tests such as door ajar alarm and temperature monitoring
  • Temperature mapping covers open door recovery and power failure simulations
  • Performed by a qualified engineer using calibrated data loggers
  • Detailed reports provided for validation and compliance
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific Liquid Nitrogen Storage Equipment
Product Type IQ/OQ Service