missing translation for 'onlineSavingsMsg'
Learn More

IPA-3, MedChemExpress

Product Code. 16499277
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499277 5 mg 5mg
16486728 10 mg 10mg
16402817 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16499277 Supplier Medchem Express Supplier No. HY15663
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress IPA-3 is a selective non-ATP competitive PAK1 inhibitor with IC50 of 2.5 μM, and shows no inhibition to group II PAKs (PAKs 4-6).

IPA-3 is a selective non-ATP competitive PAK1 inhibitor with IC50 of 2.5 μM, and shows no inhibition to group II PAKs (PAKs 4-6).

Specifications

Chemical Name or Material IPA-3
CAS 42521-82-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H14O2S2
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (285.35 mM)
SMILES OC1=CC=C2C=CC=CC2=C1SSC3=C4C=CC=CC4=CC=C3O
Molecular Weight (g/mol) 350.45
Formula Weight 350.45
Percent Purity 98.16%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.