missing translation for 'onlineSavingsMsg'
Learn More

Ipatasertib, MedChemExpress

Product Code. 16491756
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491756 500 mg 500mg
16486348 10 mg 10mg
16478937 5 mg 5mg
16461166 200 mg 200mg
16422467 50 mg 50mg
16411489 1 g 1g
16486656 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16491756 Supplier Medchem Express Supplier No. HY15186
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ipatasertib (GDC-0068) is a highly selective and ATP-competitive pan-Akt inhibitor with IC50s of 5, 18 and 8 nM for Akt1, Akt2 and Akt3, respectively.

Ipatasertib (GDC-0068) is a highly selective and ATP-competitive pan-Akt inhibitor with IC50s of 5, 18 and 8 nM for Akt1, Akt2 and Akt3, respectively.

Specifications

Chemical Name or Material Ipatasertib
Physical Form Powder
CAS 1001264-89-6
Color Off-White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C24H32ClN5O2
For Use With (Application) Cancer-Kinase/protease
Synonym GDC-0068 RG7440
Solubility Information DMSO : 220 mg/mL (480.35 mM; Need ultrasonic)
SMILES ClC1=CC=C([C@@H](CNC(C)C)C(N2CCN(C3=C([C@H](C)C[C@H]4O)C4=NC=N3)CC2)=O)C=C1
Molecular Weight (g/mol) 458.0
Formula Weight 458.0
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.