Learn More
Memmert™ IQ Document with Test Data for UN160plus
Product Code. 17599905 Shop All Memmert Products
Click to view available options
:
For Use With: UN160plus
Product Code. 17599905
Brand: Memmert™ UN160plusD00124
IQ document with device-specific works test data for validation by customer.
  • OQ/PQ check list as support for validation by customer
Specifications
For Use With (Equipment) UN160plus Oven
Product Type IQ Document
Includes Test Data