missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ IQ Document with Test Data for UN30plus

Product Code. 17570116 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
UN30plus Oven
Packungsgröße:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. For Use With (Equipment) Packungsgröße
17570116 UN30plus Oven Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17570116 Supplier Memmert™ Supplier No. UN30plusD00124
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

IQ document with device-specific works test data for validation by customer.

  • OQ/PQ check list as support for validation by customer

Specifications

For Use With (Equipment) UN30plus Oven
Product Type IQ Document
Includes Test Data
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.