Learn More
Memmert™ Iq Document With Device-Specific Works Test Data For Validation By Customer, For Icp260
Product Code. 16307027 Shop All Memmert Products
Product Code. 16307027
Brand: Memmert™ ICP260D00124
Includes
Works test data
Includes IQ document with works test data for chamber.
  • OQ/PQ check list as support for validation by customer
Specifications
For Use With (Equipment) 30, 55, 75, 110, 160, 260, 450, 750, 1060 Models Climate Chamber
Product Type IQ Document