Learn More
Memmert™ Iq/Oq Document W/Device-Specific Works Test Data (Temperature And Humidity Value), For Hcp150
Includes temperature distribution survey for one free-selectable temperature and humidity value.
Brand:  Memmert™ HCP150D00136
Additional Details : Weight : 0.01100kg
Product Code. 16374817
  • 3200.00 € / Each
Estimated Shipment: 20-06-2024
to see stock.
Includes: Works test data, temperature distribution survey
Description
Description
  • IQ/OQ document with works test data for chamber
  • 27 measuring points to DIN 12880:2007-05, PQ check list as support for validation by customer (models HPP and ICH)
Specifications
Specifications
110, 260, 750 Models Constant Climate Chamber
IQ/OQ Documentation
One Free-Selectable Temperature and Humidity Value for 27 Measuring Points to DIN 12880:2007-05