Learn More
Grant Instruments™ IQ/OQ Documentation
Product Code. 15480474
Change view
Click to view available options
For Use With (Equipment):
All Grant BTD Block Heater Models
All Grant XUBA Analogue Ultrasonic Baths
All QBD Block Heater Models
All QBH Block Heater Models
All SBB Aqua Plus Models
All SUB Aqua Plus Models
CS200G Stirred Thermostatic Bath/Circulator
CXG Stirred Thermostatic Bath/Circulator
Grant CVP-2 PCR Plate Centrifuge
Grant PHMP-100 Thermoshaker for Microplates
Grant R4 Refrigerated Baths/Circulators
Grant T100 Stirred Thermostatic Bath/Circulator
Grant TC120 Stirred Thermostatic Bath/Circulator
Grant TS-DW Deep Well Plate Thermoshaker
Grant TX150 Stirred Thermostatic Bath/Circulator
Grant TXF200 Stirred Thermostatic Bath/Circulator
JBA and SBB Unstirred Baths
JBN/JBA Baths
Unit Size:
Each
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15480474 Grant TS-DW Deep Well Plate Thermoshaker Each
15271186 All QBH Block Heater Models Each
15211196 Grant R4 Refrigerated Baths/Circulators Each
15221196 JBA and SBB Unstirred Baths Each
15251196 All SUB Aqua Plus Models Each
15211186 All Grant BTD Block Heater Models Each
15261186 All QBD Block Heater Models Each
15231186 JBN/JBA Baths Each
15241196 All SBB Aqua Plus Models Each
15281196 Grant TX150 Stirred Thermostatic Bath/Circulator Each
15201206 All Grant XUBA Analogue Ultrasonic Baths Each
15102625 CS200G Stirred Thermostatic Bath/Circulator Each
15182615 CXG Stirred Thermostatic Bath/Circulator Each
15122615 Grant T100 Stirred Thermostatic Bath/Circulator Each
15142615 Grant TC120 Stirred Thermostatic Bath/Circulator Each
15162615 Grant TXF200 Stirred Thermostatic Bath/Circulator Each
15410484 Grant CVP-2 PCR Plate Centrifuge Each
15440484 Grant PHMP-100 Thermoshaker for Microplates Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Product Code. 15480474 Supplier Grant Instruments™ Supplier No. IQOQ TSDW
  • ONLINE EXCLUSIVE PRICE
    238.00 €
    206.51 € / Pack of 1
    Save 31.49 €
    13% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Offers equipment user greater comparability and verification with their instruments. Grant Instruments™ IQ/OQ Documentation simplifies installation and maintenance procedures, reduces common user errors, and builds a clear record of an instrument's performance throughout its life.
  • Installation Qualification (IQ): instrument has been delivered and installed in accordance with manufacturer's requirements
  • Operational Qualification (OQ): instrument is functioning in accordance with its specification
  • Each IQ/OQ documentation set is specific to individual serial numbered equipment and is provided in an easy to use ring bound format.
  • It is recommend to purchase IQ/OQ documentation at the time of ordering Grant Scientific equipment.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Grant TS-DW Deep Well Plate Thermoshaker
Product Type IQ/OQ Documentation