Learn More
IQGAP1 Monoclonal Antibody (OTI6E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809921S
Encompass_Preferred
 View more versions of this product
Product Code. 17582574
  • 242.00 € / 30µL
Description
IQGAP1 Monoclonal antibody specifically detects IQGAP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
IQGAP1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P37837
Mouse
Affinity chromatography
RUO
6888
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI6E1
Unconjugated
TALDO1
AA682088; Cdc42-Rac1 effector protein; D7Ertd237e; D7Ertd257e; DISP2; DISPB; HUMORFA01; IQ motif containing GTPase activating protein 1; IQGAP1; KIAA0051; mKIAA0051; p195; Ras GTPase-activating-like protein IQGAP1; RasGAP-like with IQ motifs; SAR1
Full length human recombinant protein of human TALDO1 produced in HEK293T cell.
30 μL