missing translation for 'onlineSavingsMsg'
Learn More

IQGAP1 Mouse, anti-Human, Clone: 2E11A2, Proteintech

Product Code. 18005025 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18005025 150 μL 150µL
18005515 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18005025 Supplier Proteintech Supplier No. 685881IG150UL
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The IQGAP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IQGAP1. This antibody recognizes human, rat antigen. The IQGAP1 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen IQGAP1
Applications Western Blot, ELISA
Clone 2E11A2
Concentration 500 μg/mL
Gene Symbols HUMORFA01, IQGAP1, KIAA0051, p195, SAR1
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 8826
Target Species Human, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.