missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IQGAP2 Polyclonal Antibody
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Binds to activated CDC42 and RAC1 but does not seem to stimulate their GTPase activity. Associates with calmodulin.
Specifications
Specifications
| Antigen | IQGAP2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with trehalose and no preservative |
| Gene | IQGAP2 |
| Gene Accession No. | Q13576 |
| Gene Alias | 4933417J23Rik; A630053O10; AI788777; IQ motif containing GTPase activating protein 2; Iqgap2; Ras GTPase-activating-like protein IQGAP2 |
| Gene Symbols | IQGAP2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?