Learn More
Unity Lab Services™ IQOQ Single Unit Co2 Incubators 3rd Party
Product Code. 30286564
Change view
Click to view available options
For Use With (Equipment):
CO2 Incubators
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
30286564 CO2 Incubators Each
1 options
Product Code. 30286564 Supplier Unity Lab Services™ Supplier No. IOQCO2MVS
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Installation and operational qualification service of CO2 incubator equipment not manufactured by Thermo Fisher Scientific. Ensures equipment meets specs and functions properly, key tests like door ajar and temperature.
  • Installation and operational qualification service of CO2 incubator equipment manufactured by third parties.
  • This service will confirm equipment is operating to defined specifications.
  • Operational qualification includes a test of temperature distribution with 9 points in the work volume.
  • The service includes a test of temperature distribution at one temperature setpoint.
  • NOTE! For the support of Memmert products, validation products must be ordered separately under item code 16344567
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) CO2 Incubators
Product Type IQ/OQ Service