Learn More
Thermo Scientific IQOQPQ Package for ULT and CRYO -150°C to -50°C
Product Code. 32176307 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific ULT Freezers
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
32176307 Thermo Scientific ULT Freezers Each
1 options
Product Code. 32176307 Supplier Thermo Scientific Supplier No. IQOQPCKULTMVS
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provides qualification execution/support for compressor-based ULT and cryogenic freezers, including units where LN2 backup is secondary to the primary cooling system.
  • Not suitable for liquid nitrogen Dewar-type cryopreservation systems
  • Confirm whether documents are included/supplied free of charge or require separate document-pack logic
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific ULT Freezers
Product Type IQ/OQ/PQ with Completion of Documents