missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IRAK-M Recombinant Rabbit Monoclonal Antibody (23GB875), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
207.00 € - 562.00 €
Specifications
| Antigen | IRAK-M |
|---|---|
| Clone | 23GB875 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089268
![]() |
Invitrogen™
MA554018 |
100 μL |
562.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089252
|
Invitrogen™
MA554019 |
20 μL |
207.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IRAK-M Recombinant Monoclonal antibody specifically detects IRAK-M in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| IRAK-M | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9Y616 | |
| 11213 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB875 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 4833428C18Rik; AI563835; ASRT5; FLJ13601; IL-1 receptor-associated kinase M; interleukin 1 receptor associated kinase 3; interleukin-1 receptor-associated kinase 3; IRAK3; IRAK-3; IRAKM; IRAK-M | |
| A synthetic peptide of human IRAK3 (550-596AA). | |
| Primary | |
| -20°C | |
| IRAK3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title