missing translation for 'onlineSavingsMsg'
Learn More

Irbesartan, MedChemExpress

Product Code. 16492056
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492056 500 mg 500mg
16481396 200 mg 200mg
16474297 50 mg 50mg
16408418 10 mg 10mg
16408176 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16492056 Supplier Medchem Express Supplier No. HYB0202
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Irbesartan (SR-47436) is an orally active Ang II type 1 (AT1) receptor blocker (ARB). Irbesartan can relax the blood vessels, low blood pressure and increase the supply of blood and oxygen to the heart. Irbesartan can be used for the research of high blood pressure, heart failure, and diabetic kidney disease.

Irbesartan (SR-47436) is an orally active Ang II type 1 (AT1) receptor blocker (ARB). Irbesartan can relax the blood vessels, low blood pressure and increase the supply of blood and oxygen to the heart. Irbesartan can be used for the research of high blood pressure, heart failure, and diabetic kidney disease.

Specifications

Chemical Name or Material Irbesartan
Physical Form Solid
CAS 138402-11-6
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C25H28N6O
For Use With (Application) COVID-19-immunoregulation
Synonym SR-47436 BMS-186295
Solubility Information DMSO : 100 mg/mL (233.36 mM; Need ultrasonic)
SMILES O=C1N(CC2=CC=C(C3=C(C4=NN=NN4)C=CC=C3)C=C2)C(CCCC)=NC15CCCC5
Molecular Weight (g/mol) 428.53
Formula Weight 428.53
Percent Purity 98.98%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.