missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRE1 alpha, p Ser724 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 252 publications
311.00 € - 624.00 €
Specifications
| Antigen | IRE1 alpha (p Ser724) |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Immunoblotting, In vitro assay, Chromatin Immunoprecipitation (ChIP), KnockDown Validated |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18730434
|
Novus Biologicals
NB100-2323SS |
0.025 mL |
311.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18707033
|
Novus Biologicals
NB100-2323 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IRE1 alpha Polyclonal specifically detects IRE1 alpha in Human, Mouse, Rat, Porcine, Drosophila, Goat, Mammal, Monkey, Primate, Rabbit, Golden Syrian Hamster samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, In vitro assay, Knockdown Validated.Specifications
| IRE1 alpha (p Ser724) | |
| Western Blot 1:500 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Immunoblotting, In vitro assay, Chromatin Immunoprecipitation (ChIP), KnockDown Validated | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Neuroscience, Phospho Specific, Signal Transduction, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
| PBS with 0.02% Sodium Azide | |
| EC 2.7.11, endoplasmic reticulum to nucleus signaling 1, Endoplasmic reticulum-to-nucleus signaling 1, ER to nucleus signalling 1, FLJ30999, hIRE1p, inositol-requiring 1, Inositol-requiring protein 1, IRE 1a, ire1 alpha p Ser724, ire1 alpha phosphorylation, IRE1a, ire1-alpha, IRE1MGC163279, IRE1P, MGC163277, phospho ire1 alpha, phospho-ire1 alpha, phospho-ire1a, p-ire1 alpha, protein kinase/endoribonuclease, serine/threonine-protein kinase/endoribonuclease IRE1 | |
| ERN1 | |
| IgG | |
| Affinity Purified | |
| NB100-2323 IRE1 alpha [p Ser724] Antibody detects IRE-1 alpha when phosphorylated at Ser724 residue. |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig, Drosophila, Goat, Mammalia, Monkey, Primate, Rabbit, Golden Syrian Hamster | |
| O75460 | |
| 2081 | |
| This IRE1 alpha [p Ser724] antibody was raised against a synthetic peptide surrounding the phosphorylated serine 724 of the human IRE1 alpha protein. [Swiss-Prot #O75460] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 110 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title