Learn More
Invitrogen™ IREB2 Recombinant Rabbit Monoclonal Antibody (1Q7R2)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558031
Description
Immunogen sequence: SLEAPDSVLF PWDLKSTYIR CPSFFDKLTK EPIALQAIEN AHVLLYLGDS VTTDHISPAG SIARNSAAAK YLTNRGLTPR EFNSYGARRG NDAVMTRGTF ANIKLFNKFI GKPAPKTIHF PSGQTLDVFE AAELYQKEGI PLIILAGKKY GSGNSRDWAA KGPYLLGVKA VLAESYEKIH KDHLIGIGIA PLQFLPGENA DSLGLSGRET FSLTFPEELS PGITLNIQTS TGKVFSVIAS FEDDVEITLY KHGGLLNFVA RKFS.
Iron regulatory protein 2 (IRP2) is a central post-transcriptional regulator of cellular and systemic iron metabolism. It is a cytosolic RNA-binding protein that binds to stem-loop structures. IRP2 is regulated by iron-mediated proteolysis. Hypoxia is important for normal tissue physiology as well as being a component of several pathophysiological conditions, including heart and cerebrovascular diseases and tumor growth. Because iron and oxygen are closely related, IRP2 also plays a role during hypoxia (low oxygen).
Specifications
| IREB2 | |
| Recombinant Monoclonal | |
| 0.25 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P48200, Q62751, Q811J3 | |
| IREB2 | |
| Recombinant protein (or fragment). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1Q7R2 | |
| Unconjugated | |
| IREB2 | |
| ACO3; D9Ertd85e; FLJ23381; im:7153062; Ireb2; IRE-BP 2; IREBP2; Iron regulatory protein 2; iron responsive element binding protein 2; iron-regulatory protein 2; iron-responsive element binding protein 2; iron-responsive element-binding protein 2; Iron-responsive element-binding protein 2-like protein; IRP2; IRP2AD | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3658, 64602, 64831 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.