Learn More
IRF4 Antibody (CL14615) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26863JF549
Product Code. 30305366
  • 770.00 € / 0.10mL
Description
IRF4 Monoclonal antibody specifically detects IRF4 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IRF4
Monoclonal
Janelia Fluor 549
interferon regulatory factor 4, IRF-4, LSIRFMUM1, Lymphocyte-specific interferon regulatory factor, Multiple myeloma oncogene 1, NF-EM5
This antibody was developed using a recombinant protein derived from Q15306, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
3662
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot
CL14615
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified