missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99072-100ul
This item is not returnable.
View return policy
Description
IRF6 Polyclonal antibody specifically detects IRF6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| IRF6 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| interferon regulatory factor 6, IRF-6, LPS, OFC6PPS, PIT, Van der Woude syndrome, VWS, VWS1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human IRF6. | |
| 100 μg | |
| Immunology, Innate Immunity, Stem Cells | |
| 3664 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction