missing translation for 'onlineSavingsMsg'
Learn More

Irinotecan hydrochloride, MedChemExpress

Product Code. 16487116
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487116 100 mg 100mg
16483117 50 mg 50mg
16431226 200 mg 200mg
16401816 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16487116 Supplier Medchem Express Supplier No. HY16562A
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Irinotecan hydrochloride ((+)-Irinotecan hydrochloride) is a topoisomerase I inhibitor mainly used to treat colon cancer and rectal cancer.

Irinotecan hydrochloride ((+)-Irinotecan hydrochloride) is a topoisomerase I inhibitor mainly used to treat colon cancer and rectal cancer.

Specifications

Chemical Name or Material Irinotecan hydrochloride
CAS 100286-90-6
Color White
Purity Grade Notes Research
Molecular Formula C33H39ClN4O6
Quantity 100 mg
Synonym (+)-Irinotecan hydrochlorideCPT-11 hydrochloride
Solubility Information DMSO : 125 mg/mL (200.60 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (5.34 mM; Need ultrasonic)
SMILES O=C(N1CCC(N2CCCCC2)CC1)OC3=CC=C4N=C5C(CN6C(C(COC([C@@]7(CC)O)=O)=C7C=C65)=O)=C(CC)C4=C3.[H]Cl
Molecular Weight (g/mol) 623.14
Formula Weight 623.14
Percent Purity 98.03%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.